Learn More
Electron Microscopy Sciences Veco Hexagonal with Handle 400 mesh Nickel Grid
SDP
Catalog No. 5026430
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Nickel
Pitch: 63 µm
Hole: 30 µm
Bar: 33 µm
Catalog No. 50-264-30 Supplier Electron Microscopy Sciences Supplier No. HD400HNI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Grid: Nickel
Pitch: 63 µm
Hole: 30 µm
Bar: 33 µm