Learn More
Grainger Hex Head Cap, Brass
Catalog No. NC2455883
Encompass_Preferred
The Hex Head Cap Brass Screw is a brass screw with a hexagonal head. It has a thread diameter of 1/4"-20 and a length of 1". The screw has a head height of 0.281" and width across flats of 0.5". It is made from 360 brass and meets ASME B18.2.1 specifications. The screw has a tensile strength of 61,000 psi. It can be used for various fastening applications.
Catalog No. NC2455883 Supplier Grainger Supplier No. 6AYZ3
The Hex Head Cap Brass Screw is a brass screw with a hexagonal head. It has a thread diameter of 1/4"-20 and a length of 1". The screw has a head height of 0.281" and width across flats of 0.5". It is made from 360 brass and meets ASME B18.2.1 specifications. The screw has a tensile strength of 61,000 psi. It can be used for various fastening applications.