Learn More
Grainger Hex Head Plug 316L SS 3/4 in MNPT
Catalog No. 502752775
Encompass_Preferred
The Parker Hex Head Plug is made from 316L stainless steel. It has a hex head design and is available in 1/4", 3/8", and 1/2" sizes. The plugs have a pressure rating of up to 10,000 psi and temperature rating of up to 850°F. They are designed for use in hydraulic and pneumatic systems to seal fluid ports. The hex head allows for easy installation and removal using a wrench.
Catalog No. 50-275-2775 Supplier Grainger Supplier No. 14M061
The Parker Hex Head Plug is made from 316L stainless steel. It has a hex head design and is available in 1/4", 3/8", and 1/2" sizes. The plugs have a pressure rating of up to 10,000 psi and temperature rating of up to 850°F. They are designed for use in hydraulic and pneumatic systems to seal fluid ports. The hex head allows for easy installation and removal using a wrench.