Learn More
Grainger Hex Head Plug Forged Steel 1/2 in
Catalog No. 502749534
Encompass_Preferred
The Anvil Hex Head Plug is a forged steel plug with a hex head drive. It is made in the USA and has a black oxide finish. The plug has coarse threads and is available in sizes ranging from 1/4"-20 to 1"-8. It can be used to plug holes in metal, wood, or plastic. The plugs are sold individually.
Catalog No. 50-274-9534 Supplier Grainger Supplier No. 14J749
The Anvil Hex Head Plug is a forged steel plug with a hex head drive. It is made in the USA and has a black oxide finish. The plug has coarse threads and is available in sizes ranging from 1/4"-20 to 1"-8. It can be used to plug holes in metal, wood, or plastic. The plugs are sold individually.