Learn More
Grainger Hex Head Plug Forged Steel 2 in MNPT
Catalog No. 502749536
Encompass_Preferred
The Anvil Hex Head Plug is a forged steel plug with a hex head drive for use in sealing pipe openings. It has a pressure rating of 150 psi and temperature rating of 650°F. The plug has a standard pipe thread and is available in sizes ranging from 1/8 inch to 2 inches. It is made from hot rolled and completely annealed steel for strength and machinability. The heavy hex head drive allows use with standard wrenches for installation and removal. This plug provides an economical way to temporarily or permanently seal openings in threaded pipe systems.
Catalog No. 50-274-9536 Supplier Grainger Supplier No. 14J753
The Anvil Hex Head Plug is a forged steel plug with a hex head drive for use in sealing pipe openings. It has a pressure rating of 150 psi and temperature rating of 650°F. The plug has a standard pipe thread and is available in sizes ranging from 1/8 inch to 2 inches. It is made from hot rolled and completely annealed steel for strength and machinability. The heavy hex head drive allows use with standard wrenches for installation and removal. This plug provides an economical way to temporarily or permanently seal openings in threaded pipe systems.