Learn More
Grainger Hex Head Plug Forged Steel 3/4 in
Catalog No. 502748808
Encompass_Preferred
The Hex Head Plug is a forged steel plug with a hex head drive for tightening. It has a thread size of 1/4"-20 and a length of 1". The plug is made from grade 8 steel for strength and has a zinc yellow dichromate finish for corrosion resistance. The maximum pressure rating is 3000 psi. The hex head plug can be used to seal threaded ports in machinery, engines, and hydraulic components.
Catalog No. 50-274-8808 Supplier Grainger Supplier No. 14J750
The Hex Head Plug is a forged steel plug with a hex head drive for tightening. It has a thread size of 1/4"-20 and a length of 1". The plug is made from grade 8 steel for strength and has a zinc yellow dichromate finish for corrosion resistance. The maximum pressure rating is 3000 psi. The hex head plug can be used to seal threaded ports in machinery, engines, and hydraulic components.