Learn More
Electron Microscopy Sciences Hex Key Wrenches, English Set 1
SDP
Catalog No. 50996503
Encompass_Preferred
Made from heat treated chrome nickel steel. Useful to have around the lab. Its sturdy handle, folds up just like a pocket knife. Can be used at any angle. Sizes in inches: .05, 1/16, 5/64, 3/32, 7/64, 1/8, 9/64, 5/32, 3/16. Handle length: 3-5/8".
Catalog No. 50-996-503 Supplier Electron Microscopy Sciences Supplier No. 6207501
Made from heat treated chrome nickel steel. Useful to have around the lab. Its sturdy handle, folds up just like a pocket knife. Can be used at any angle. Sizes in inches: .05, 1/16, 5/64, 3/32, 7/64, 1/8, 9/64, 5/32, 3/16. Handle length: 3-5/8".