Learn More
Grainger Hex Long Nipple 316 SS 3/4 in MNPT
Catalog No. 502752954
Encompass_Preferred
The Parker Hex Long Nipple is made from 316 stainless steel and has a hexagonal shape. It has NPT threads on both ends and is available in sizes ranging from 1/8" to 2" diameter. The nipple can be used to extend and connect threaded pipe. It meets ASME B1.20.1 standards and is rated to handle pressures up to 6000 psi at 100°F. The 316 stainless construction makes it durable and corrosion resistant. This hex nipple provides a secure threaded pipe connection.
Catalog No. 50-275-2954 Supplier Grainger Supplier No. 14M050
The Parker Hex Long Nipple is made from 316 stainless steel and has a hexagonal shape. It has NPT threads on both ends and is available in sizes ranging from 1/8" to 2" diameter. The nipple can be used to extend and connect threaded pipe. It meets ASME B1.20.1 standards and is rated to handle pressures up to 6000 psi at 100°F. The 316 stainless construction makes it durable and corrosion resistant. This hex nipple provides a secure threaded pipe connection.