Learn More
Grainger Hex Long Nipple 316 SS 3/4 in MNPT
Catalog No. 502752603
Encompass_Preferred
The Parker Hex Long Nipple is a 316 stainless steel pipe fitting with male NPT threads on both ends. It has a hexagonal body for wrench tightening and is made from 316 stainless steel for corrosion resistance. This nipple has a pressure rating of 6000 psi at 100°F and is available in sizes ranging from 1/8" to 2" NPT in lengths from 1" to 24". It can be used to extend and connect stainless steel pipe.
Catalog No. 50-275-2603 Supplier Grainger Supplier No. 14M062
The Parker Hex Long Nipple is a 316 stainless steel pipe fitting with male NPT threads on both ends. It has a hexagonal body for wrench tightening and is made from 316 stainless steel for corrosion resistance. This nipple has a pressure rating of 6000 psi at 100°F and is available in sizes ranging from 1/8" to 2" NPT in lengths from 1" to 24". It can be used to extend and connect stainless steel pipe.