Learn More
Grainger Hex Long Nipple 316 SS 3/4 x 1/2 in
Catalog No. 502751907
Encompass_Preferred
The Parker Hex Long Nipple is made from 316 stainless steel and has a hexagonal shape. It has NPT threads on both ends and is available in sizes ranging from 1/8" to 2" diameter. The nipple can be used to extend and connect threaded pipe. It meets ASTM A269 standards. The 316 stainless construction offers corrosion resistance and durability. The hex shape allows for easy tightening with a wrench. This nipple provides a high quality threaded pipe connection.
Catalog No. 50-275-1907 Supplier Grainger Supplier No. 14M049
The Parker Hex Long Nipple is made from 316 stainless steel and has a hexagonal shape. It has NPT threads on both ends and is available in sizes ranging from 1/8" to 2" diameter. The nipple can be used to extend and connect threaded pipe. It meets ASTM A269 standards. The 316 stainless construction offers corrosion resistance and durability. The hex shape allows for easy tightening with a wrench. This nipple provides a high quality threaded pipe connection.