Learn More
Grainger Hex Long Nipple 316 SS 3/4 x 1/4 in
Catalog No. 502752602
Encompass_Preferred
The Parker Hex Long Nipple is made from 316 stainless steel. It has a hexagonal shape with a 1/2" NPT thread size on both ends. The overall length is 3" and it has a schedule 40 wall thickness. This nipple can be used to extend and connect threaded pipe fittings. The 316 stainless construction offers strength and corrosion resistance. With a maximum pressure rating of 6000 psi at 100°F, this nipple is well-suited for high pressure hydraulic applications. The hexagonal shape allows for easy tightening with a wrench.
Catalog No. 50-275-2602 Supplier Grainger Supplier No. 14M048
The Parker Hex Long Nipple is made from 316 stainless steel. It has a hexagonal shape with a 1/2" NPT thread size on both ends. The overall length is 3" and it has a schedule 40 wall thickness. This nipple can be used to extend and connect threaded pipe fittings. The 316 stainless construction offers strength and corrosion resistance. With a maximum pressure rating of 6000 psi at 100°F, this nipple is well-suited for high pressure hydraulic applications. The hexagonal shape allows for easy tightening with a wrench.