Learn More
Electron Microscopy Sciences Gilder Hexagonal 75 mesh Nickel Grid
SDP
Catalog No. 501901235
Encompass
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. The copper grids with palladium plating offers better grid strength and avoids tarnishing. 3.05 mm diameter, 0.7 mil (18um) thickness.
Grid: Nickel
Pitch: 340 µm
Hole: 290 µm
Bar: 50 µm
Catalog No. 50-190-1235 Supplier Electron Microscopy Sciences Supplier No. G75HNI
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. The copper grids with palladium plating offers better grid strength and avoids tarnishing. 3.05 mm diameter, 0.7 mil (18um) thickness.
Grid: Nickel
Pitch: 340 µm
Hole: 290 µm
Bar: 50 µm