Learn More
AdipoGen Hex1 mAb recSH1237-F8
SDP
Supplier:  AdipoGen AG27B6324C050
Encompass_Preferred
Recombinant Antibody. Recognizes Hex1 from Neurospora crassa. Applications ELISA, IGS. Clone SH1237-F8. Isotype Human IgG1. Formulation Liquid. In PBS. Neurospora crassa has been an excellent model organism for eukaryotic genetics and biochemistry and one of the workhorses for fungal cell biology research. The rapid mm~4 hr/1 filamentous growth habit of N. crassa is the result of a strongly polarized mechanism culminating in the biogenesis of the tubular cell wall. Woronin bodies WBsperoxisome-related are membrane-bound organelles of filamentous ascomycetes that mediate hyphal compartmentalization by plugging septal pores upon hyphal damage. Their major component is the peroxisomal protein HEX-1, which has also been implicated in additional cellular processes in fungi.
Catalog No. 50-256-3119