Learn More
Aviva Systems Biology Corp HEXA Antibody - C-terminal region (ARP61100_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61100_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HEXA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HEXA. Alternate names: MGC99608, TSDApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-287-251