Learn More
Selleck Chemical LLC Hexa-D-arginine-E0029-25MG
SDP
Catalog No. 503135716
Encompass_Preferred
Hexa-D-arginine is a potent and specific furin inhibitor with Ki values of 106 nM 580 nM and 13 2 M for furin PACE4 and PC1 respectively
Catalog No. 50-313-5716 Supplier Selleck Chemical LLC Supplier No. E002925MG
Hexa-D-arginine is a potent and specific furin inhibitor with Ki values of 106 nM 580 nM and 13 2 M for furin PACE4 and PC1 respectively