Learn More
Medchemexpress LLC Hexa-D-arginine (Furin Inhibitor II) | 673202-67-0 | 100.0% | 954.14 | 10 MG
SDP
Catalog No. 5000426230
Encompass_Preferred
Hexa-D-arginine (Furin Inhibitor II) is a stable furin inhibitor with Ki values of 106 nM for furin, 580 nM for PACE4, and 13.2 μM for prohormone convertase-1 (PC1). It effectively blocks Pseudomonas exotoxin A and anthrax toxins toxicity both in vitro and in vivo. Administration significantly improves survival rate in PEA-treated mice and decreases circulating levels of TNF-α.
  • Stable furin inhibitor
  • Blocks Pseudomonas exotoxin A and anthrax toxins toxicity
  • Improves survival rate in PEA-treated mice
  • Decreases circulating levels of TNF-α
Catalog No. 50-004-26230 Supplier Medchemexpress LLC Supplier No. HYP102810MG
Hexa-D-arginine (Furin Inhibitor II) is a stable furin inhibitor with Ki values of 106 nM for furin, 580 nM for PACE4, and 13.2 μM for prohormone convertase-1 (PC1). It effectively blocks Pseudomonas exotoxin A and anthrax toxins toxicity both in vitro and in vivo. Administration significantly improves survival rate in PEA-treated mice and decreases circulating levels of TNF-α.
  • Stable furin inhibitor
  • Blocks Pseudomonas exotoxin A and anthrax toxins toxicity
  • Improves survival rate in PEA-treated mice
  • Decreases circulating levels of TNF-α