Learn More
Medchemexpress LLC Hexa-D-arginine TFA | 958204-56-3 | MFCD18633383 | 98.4% | 1068.17 | 5 MG
SDP
Catalog No. 5000425444
Encompass_Preferred
Hexa-D-arginine TFA (Furin Inhibitor II TFA) is a stable furin inhibitor with Ki values 106 nM, 580 nM and 13.2 μM for furin, PACE4 and prohormone convertase-1 (PC1), respectively. It blocks Pseudomonas exotoxin A and anthrax toxins toxicity in vitro and in vivo.
  • Stable furin inhibitor
  • Inhibits PACE4 and prohormone convertase-1 (PC1)
  • Blocks Pseudomonas exotoxin A toxicity
  • Blocks anthrax toxins toxicity
Catalog No. 50-004-25444 Supplier Medchemexpress LLC Supplier No. HYP1028A5MG
Hexa-D-arginine TFA (Furin Inhibitor II TFA) is a stable furin inhibitor with Ki values 106 nM, 580 nM and 13.2 μM for furin, PACE4 and prohormone convertase-1 (PC1), respectively. It blocks Pseudomonas exotoxin A and anthrax toxins toxicity in vitro and in vivo.
  • Stable furin inhibitor
  • Inhibits PACE4 and prohormone convertase-1 (PC1)
  • Blocks Pseudomonas exotoxin A toxicity
  • Blocks anthrax toxins toxicity