Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexa(ethylene glycol) dithiol | 89141-22-0 | 500 MG
SDP

Catalog No. 5000340527
Encompass_Preferred

Hexa(ethylene glycol) dithiol, also known as Thiol-PEG5-thiol (Bis-PEG5-thiol), is a bifunctional thiol cross-linker agent. It is intended for research and development applications, specifically targeting biochemical assay reagents. The chemical has a formula of C12H26O5S2 and a molecular weight of 314.46.

  • Bifunctional thiol cross-linker agent
  • Targets biochemical assay reagents
  • Chemical formula: C12H26O5S2
  • Molecular weight: 314.46
  • Relative density: 1.113 g/cm³ at 25 °C
  • Flash point: >110 °C
  • Recommended storage at 2-8 °C
  • Handle under nitrogen, protected from moisture

Catalog No. 50-003-40527 Supplier Medchemexpress LLC Supplier No. HY130601500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hexa(ethylene glycol) dithiol, also known as Thiol-PEG5-thiol (Bis-PEG5-thiol), is a bifunctional thiol cross-linker agent. It is intended for research and development applications, specifically targeting biochemical assay reagents. The chemical has a formula of C12H26O5S2 and a molecular weight of 314.46.

  • Bifunctional thiol cross-linker agent
  • Targets biochemical assay reagents
  • Chemical formula: C12H26O5S2
  • Molecular weight: 314.46
  • Relative density: 1.113 g/cm³ at 25 °C
  • Flash point: >110 °C
  • Recommended storage at 2-8 °C
  • Handle under nitrogen, protected from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.