Learn More
Strem, An Ascensus Company CAS# 813-19-4. 10g. Hexa-n-butylditin, min. 95%. MFCD00009417
SDP
Supplier:  Strem, An Ascensus Company 93501910G
Encompass
CAS# 813-19-4. 10g. Hexa-n-butylditin, min. 95%. MFCD00009417. Molecular Weight: 580.08. Molecular Formula: (C4H9)3SnSn(C4H9)3. Color/form: colorless to yellow liq. Strem# 93-5019. http://www.strem.com/catalog/v/93-5019/
Catalog No. 50-901-15566