Learn More
Strem, An Ascensus Company CAS# 10534-89-1. 10g. Hexaamminecobalt(III) chloride (99.999%-Co) PURATREM. MFCD00036304
SDP
Catalog No. 5090114716
Encompass
CAS# 10534-89-1. 10g. Hexaamminecobalt(III) chloride (99.999%-Co) PURATREM. MFCD00036304. Molecular Weight: 267.46. Molecular Formula: [Co(NH3)6]Cl3. Color/form: orange pwdr. Strem# 93-2708. http://www.strem.com/catalog/v/93-2708/
Catalog No. 50-901-14716 Supplier Strem, An Ascensus Company Supplier No. 93270810G
CAS# 10534-89-1. 10g. Hexaamminecobalt(III) chloride (99.999%-Co) PURATREM. MFCD00036304. Molecular Weight: 267.46. Molecular Formula: [Co(NH3)6]Cl3. Color/form: orange pwdr. Strem# 93-2708. http://www.strem.com/catalog/v/93-2708/