Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexaamminecobalt(III) chloride | 10534-89-1 | 99.5% | 267.48 g/mol | Cl3CoH18N6 | 25 G
SDP

Catalog No. 5000208928
Encompass_Preferred

Hexaamminecobalt(III) chloride is a cobalt coordination compound (CAS 10534-89-1) with a molecular weight of 267.48 g/mol and a stated purity of 99.5%. It is supplied as a solid for research applications and is commonly provided in small pack sizes suitable for laboratory use.

  • High stated purity of 99.5%.
  • Solid cobalt coordination complex with formula Cl3CoH18N6.
  • Molecular weight 267.48 g/mol.
  • CAS number 10534-89-1 for chemical identification.
  • Available in small research pack sizes, including 25 g.

Catalog No. 50-002-08928 Supplier Medchemexpress LLC Supplier No. 25GHYW075903
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexaamminecobalt(III) chloride is a cobalt coordination compound (CAS 10534-89-1) with a molecular weight of 267.48 g/mol and a stated purity of 99.5%. It is supplied as a solid for research applications and is commonly provided in small pack sizes suitable for laboratory use.

  • High stated purity of 99.5%.
  • Solid cobalt coordination complex with formula Cl3CoH18N6.
  • Molecular weight 267.48 g/mol.
  • CAS number 10534-89-1 for chemical identification.
  • Available in small research pack sizes, including 25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.