Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexaamminecobalt(III) chloride, 97%

Catalog No. p-4407411
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1376914 25 g
AA1376922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1376914 Supplier Thermo Scientific Chemicals Supplier No. 01376914
Only null left
Add to Cart

CAS: 10534-89-1 | [Co(NH3)6]Cl3

Hexaamminecobalt(III) chloride is used in X-ray crystallography and NMR. It is useful in DNA condensation studies. It is suitable for induction of transitions of DNA from B to A or Z forms, induction of nucleic acid crystal growth, stabilization of tertiary tRNA interactions, preparation of ligation buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10534-89-1
Molecular Formula [Co(NH3)6]Cl3
MDL Number MFCD00036304

Specifications

Color Orange
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 97%
Solubility Information Soluble in ammonia.
Formula Weight 267.48
Odor Odorless
Appearance Orange powder
Density 1.71 g/cm3
Chemical Name or Material Hexaamminecobalt(III) chloride
Hazard Category H317-H334-H350
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 234-103-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.