Learn More
Strem, An Ascensus Company CAS# 10534-88-0. 25g. Hexaamminenickel(II) chloride, 98%. MFCD00011505
SDP
Catalog No. 5090114791
Encompass
CAS# 10534-88-0. 25g. Hexaamminenickel(II) chloride, 98%. MFCD00011505. Molecular Weight: 231.80. Molecular Formula: [Ni(NH3)6]Cl2. Color/form: purple xtl. Strem# 93-2823. http://www.strem.com/catalog/v/93-2823/
Catalog No. 50-901-14791 Supplier Strem, An Ascensus Company Supplier No. 93282325G
CAS# 10534-88-0. 25g. Hexaamminenickel(II) chloride, 98%. MFCD00011505. Molecular Weight: 231.80. Molecular Formula: [Ni(NH3)6]Cl2. Color/form: purple xtl. Strem# 93-2823. http://www.strem.com/catalog/v/93-2823/