Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexaammineruthenium(II) chloride | 15305-72-3 | MFCD00011477 | 98.0% | 274.16 g/mol | [Ru(NH3)6]Cl2 | 250 MG
SDP

Catalog No. 5000303426
Encompass_Preferred

Hexaammineruthenium(II) chloride is a yellow crystalline coordination compound (formula [Ru(NH3)6]Cl2, CAS 15305-72-3) used as a research reagent in catalysis, electrochemistry, and analytical chemistry. It is supplied in small laboratory quantities for bench-scale experiments, characterization, and redox studies.

  • Coordination compound with formula [Ru(NH3)6]Cl2.
  • CAS number 15305-72-3.
  • Yellow crystalline solid, limited solubility in water.
  • Commonly used as a catalyst and redox reagent in electrochemical studies.
  • Available in small pack sizes suitable for research (e.g., 25 mg-1 g).
  • Purity commonly offered around 98% (supplier dependent).
  • Handle with appropriate safety precautions and consult the SDS before use.

Catalog No. 50-003-03426 Supplier Medchemexpress LLC Supplier No. 250MGHYW033573
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexaammineruthenium(II) chloride is a yellow crystalline coordination compound (formula [Ru(NH3)6]Cl2, CAS 15305-72-3) used as a research reagent in catalysis, electrochemistry, and analytical chemistry. It is supplied in small laboratory quantities for bench-scale experiments, characterization, and redox studies.

  • Coordination compound with formula [Ru(NH3)6]Cl2.
  • CAS number 15305-72-3.
  • Yellow crystalline solid, limited solubility in water.
  • Commonly used as a catalyst and redox reagent in electrochemical studies.
  • Available in small pack sizes suitable for research (e.g., 25 mg-1 g).
  • Purity commonly offered around 98% (supplier dependent).
  • Handle with appropriate safety precautions and consult the SDS before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.