Learn More
Strem, An Ascensus Company CAS# 14282-91-8. 5g. Hexaammineruthenium(III) chloride, 99%. MFCD00011478
SDP
Supplier:  Strem, An Ascensus Company 4406205G
Encompass
CAS# 14282-91-8. 5g. Hexaammineruthenium(III) chloride, 99%. MFCD00011478. Molecular Weight: 309.61. Molecular Formula: Ru(NH3)6Cl3. Color/form: pale yellow pwdr. Strem# 44-0620. http://www.strem.com/catalog/v/44-0620/
Catalog No. 50-901-11961