Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-α-Hexabromocyclododecane, ≥98.0% (HPLC); 99.0-101.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111018185 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8185 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8185 Supplier MilliporeSigma Supelco Supplier No. 3037325MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 134237-50-6
Linear Formula C12H18 Br6
MDL Number MFCD28118917
Synonym alpha-HBCD; (1 RS,2 RS,5 SR,6 RS,9 RS,10 SR)-1,2,5,6,9,10-Hexabromocyclododecane
Type Analytical Standard
Quantity 25 mg
Formula Weight 641.70
Percent Purity ≥98.0% (HPLC); 99.0 to 101.0% (AT)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.