Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Hexachloroacetone, 99%
Click to view available options
Quantity:
250g
1000g
Specifications
Density | 1.748 |
Quantity | 250g |
Refractive Index | 1.5115 |
Melting Point | -2°C |
Boiling Point | 203°C to 205°C |
Flash Point | >110°C (230°F) |
UN Number | UN2661 |
Beilstein | 1707475 |
Formula Weight | 264.75 |
Percent Purity | 99% |
Show More |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: HEXACHLOROACETONE
EINECSNumber : 204-129-5
RTECSNumber : UC2100000
TSCA : Yes
Recommended Storage : Ambient temperatures