Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachloroacetone, 99%

Catalog No. p-7026924
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2325930 250 g
AAB232590B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2325930 Supplier Thermo Scientific Chemicals Supplier No. B2325930
Only null left
Add to Cart

CAS: 116-16-5 | C3Cl6O | 264.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-16-5
Molecular Formula C3Cl6O
Molecular Weight (g/mol) 264.73
MDL Number MFCD00000796
InChI Key DOJXGHGHTWFZHK-UHFFFAOYSA-N
Synonym hexachloroacetone, hexachloro-2-propanone, perchloroacetone, hexachloropropanone, kureha hca, hca weedkiller, perchloro-2-propanone, bis trichloromethyl ketone, acetone, hexachloro, 2-propanone, hexachloro
PubChem CID 8303
ChEBI CHEBI:82243
IUPAC Name 1,1,1,3,3,3-hexachloropropan-2-one
SMILES ClC(Cl)(Cl)C(=O)C(Cl)(Cl)Cl

Specifications

Melting Point -2°C
Density 1.748
Boiling Point 203°C to 205°C
Flash Point >110°C (230°F)
Refractive Index 1.5115
Quantity 250 g
UN Number UN2661
Beilstein 1707475
Formula Weight 264.75
Percent Purity 99%
Chemical Name or Material Hexachloroacetone
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: HEXACHLOROACETONE
EINECSNumber 204-129-5
RTECSNumber UC2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.