Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachlorobenzene 99%, Thermo Scientific™

Catalog No. AC173885000
Change view
Click to view available options
Quantity:
10 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC173885000 500 g
AC173880100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173885000 Supplier Thermo Scientific Chemicals Supplier No. 173885000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-74-1
Molecular Formula C6Cl6
Molecular Weight (g/mol) 284.77
MDL Number MFCD00000540
InChI Key CKAPSXZOOQJIBF-UHFFFAOYSA-N
Synonym hexachlorobenzene, perchlorobenzene, phenyl perchloryl, anticarie, benzene, hexachloro, sanocide, hexachlorbenzol, snieciotox, amatin, bunt-cure
PubChem CID 8370
ChEBI CHEBI:5692
IUPAC Name hexachlorobenzene
SMILES ClC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl

Specifications

Melting Point 228°C to 231°C
Color White to Beige
Quantity 500 g
Infrared Spectrum Authentic
Physical Form Powder
Linear Formula C6Cl6
Beilstein 05, 205
Merck Index 15, 4713
Formula Weight 284.77
Percent Purity ≥98.5%
Chemical Name or Material Hexachlorobenzene, 99%
Show More Show Less
EINECSNumber 204-273-9
RTECSNumber DA2975000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.