Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachlorobenzene Standard, 100 μg/mL, Quantitative, ISO 17034, 1 mL

Catalog No. USCH1501
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-CH-150-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-CH-150-1 Supplier Agilent Technologies Supplier No. CH1501
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 118-74-1 , 75-09-2
Molecular Formula C6Cl6
Molecular Weight (g/mol) 284.77
MDL Number MFCD00000540
InChI Key CKAPSXZOOQJIBF-UHFFFAOYSA-N
PubChem CID 8370
ChEBI CHEBI:5692
IUPAC Name hexachlorobenzene
SMILES ClC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl

Specifications

CAS Max % 0.99, 0.01
Analyte or Component Names methylene chloride (dichloromethane)
Element Methylene Chloride
Solvent or Matrix Methylene Chloride
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Hexachlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.