Learn More
Cayman Chemical Hexachlorophene, Purity: ≥98%, CAS: 70-30-4, 100 mg
SDP
Catalog No. 502994423
Encompass_Preferred
An organochlorine antiseptic and antibacterial biocide; active against S. aureus (MIC = 1.56 μg/ml), E. faecium isolates from fresh produce (MICs = 5-50 mg/l), Salmonella isolates from meats (MICs = 2.5-25 mg/l), and Pseudomonas isolates from slaughterhouse surfaces (MICs = ≤0.0025-≤0.25 mg/l); inhibits infection of mouse astrocytoma DBT cells by recombinant MHV (IC50 = 1.2 μM); inhibits recombinant human glutathione transferases P1-1 and A3-3 (IC50s = 9.7 and v7 channels expressed in CHO cells (EC50 = 4.61 μM); toxic to fasted and non-fasted rats (LD50s = 215 and 165 mg/kg, respectively); reduces R. solani-induced fruit rot in cucumber plants when applied at 4.4 kg/ha
Catalog No. 50-299-4423 Supplier Cayman Chemical Supplier No. 23948100 MG
An organochlorine antiseptic and antibacterial biocide; active against S. aureus (MIC = 1.56 μg/ml), E. faecium isolates from fresh produce (MICs = 5-50 mg/l), Salmonella isolates from meats (MICs = 2.5-25 mg/l), and Pseudomonas isolates from slaughterhouse surfaces (MICs = ≤0.0025-≤0.25 mg/l); inhibits infection of mouse astrocytoma DBT cells by recombinant MHV (IC50 = 1.2 μM); inhibits recombinant human glutathione transferases P1-1 and A3-3 (IC50s = 9.7 and v7 channels expressed in CHO cells (EC50 = 4.61 μM); toxic to fasted and non-fasted rats (LD50s = 215 and 165 mg/kg, respectively); reduces R. solani-induced fruit rot in cucumber plants when applied at 4.4 kg/ha