Learn More
Cayman Chemical Hexaconazole, Purity: ≥95%, CAS: 79983-71-4, 100 mg
SDP
Catalog No. 502933280
Encompass_Preferred
A triazole fungicide; inhibits ergosterol biosynthesis via inhibition of the cytochrome P450-dependent 14α-demethylation of lanosterol; curative against powdery mildew on barley plants at 6.7 mg/L; improves water, proline, and protein contents as well as increases non-enzymatic and enzymatic antioxidant and apigenin-7-glucoside content in M. chamomilla plants during a water deficit stress tolerance test at 15 mg/L; inhibits the differentiation of mouse embryonic stem cells into cardiomyocytes (EC50 = 16.6 μM); induces bone morphological defects in mouse fetuses
Catalog No. 50-293-3280 Supplier Cayman Chemical Supplier No. 24118100 MG
A triazole fungicide; inhibits ergosterol biosynthesis via inhibition of the cytochrome P450-dependent 14α-demethylation of lanosterol; curative against powdery mildew on barley plants at 6.7 mg/L; improves water, proline, and protein contents as well as increases non-enzymatic and enzymatic antioxidant and apigenin-7-glucoside content in M. chamomilla plants during a water deficit stress tolerance test at 15 mg/L; inhibits the differentiation of mouse embryonic stem cells into cardiomyocytes (EC50 = 16.6 μM); induces bone morphological defects in mouse fetuses