Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexacosane | 630-01-3 | MFCD00009354 | 98.0% | 366.71 g/mol | C26H54 | 250 MG
SDP

Catalog No. 5000283116
Encompass_Preferred

Hexacosane (n-hexacosane) is a straight-chain alkane supplied as a purified analytical standard for laboratory research. It is suitable as an internal standard, tracer, or reference material for method development, calibration, and quantitative analysis in techniques such as GC-MS and NMR. The material is intended for research use and is accompanied by typical product documentation (COA, SDS).

  • High purity: 98.0%.
  • Molecular weight 366.71 g/mol.
  • Chemical formula C26H54.
  • Suitable as an internal standard for GC-MS and NMR.
  • Useful for method development and analytical calibration.

Catalog No. 50-002-83116 Supplier Medchemexpress LLC Supplier No. HYW131352250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hexacosane (n-hexacosane) is a straight-chain alkane supplied as a purified analytical standard for laboratory research. It is suitable as an internal standard, tracer, or reference material for method development, calibration, and quantitative analysis in techniques such as GC-MS and NMR. The material is intended for research use and is accompanied by typical product documentation (COA, SDS).

  • High purity: 98.0%.
  • Molecular weight 366.71 g/mol.
  • Chemical formula C26H54.
  • Suitable as an internal standard for GC-MS and NMR.
  • Useful for method development and analytical calibration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.