Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexacosanedioicacid | 3365-67-1 | MFCD00673440 | 99.9% | 426.67 | C26H50O4 | 250 MG
SDP

Catalog No. 5000284854
Encompass_Preferred

Hexacosanedioicacid is a long-chain dicarboxylic acid supplied as a white to off-white powder for laboratory research use. It is characterized by a molecular formula of C26H50O4, a molecular weight of 426.67, and is provided with supporting analytical documentation (datasheet, COA, SDS). The material exhibits limited solubility in common solvents and specific storage requirements to preserve stability.

  • White to off-white solid powder suitable for laboratory use.
  • High purity: 99.9% (as supplied).
  • Molecular weight 426.67 and formula C26H50O4.
  • Soluble in DMSO at approximately 2 mg/mL with ultrasonic assistance.
  • Powder storage stability: -20°C for 3 years, 4°C for 2 years.
  • In-solution stability: -80°C for 6 months, -20°C for 1 month.
  • Available in small research quantities, including 250 mg.

Catalog No. 50-002-84854 Supplier Medchemexpress LLC Supplier No. HYW613967250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hexacosanedioicacid is a long-chain dicarboxylic acid supplied as a white to off-white powder for laboratory research use. It is characterized by a molecular formula of C26H50O4, a molecular weight of 426.67, and is provided with supporting analytical documentation (datasheet, COA, SDS). The material exhibits limited solubility in common solvents and specific storage requirements to preserve stability.

  • White to off-white solid powder suitable for laboratory use.
  • High purity: 99.9% (as supplied).
  • Molecular weight 426.67 and formula C26H50O4.
  • Soluble in DMSO at approximately 2 mg/mL with ultrasonic assistance.
  • Powder storage stability: -20°C for 3 years, 4°C for 2 years.
  • In-solution stability: -80°C for 6 months, -20°C for 1 month.
  • Available in small research quantities, including 250 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.