Learn More
Medchemexpress LLC Hexacosanoic acid | 506-46-7 | 95.0% | 396.69 | 250 MG
SDP
Catalog No. 5000439886
Encompass_Preferred
Hexacosanoic acid is a very long-chain fatty acid. Abnormally elevated levels of hexacosanoic acid are closely associated with various diseases, including X-linked adrenoleukodystrophy, adrenomyeloneuropathy, atherosclerosis, and dementia.
  • Very long-chain fatty acid
  • Associated with various diseases
  • Shows antineuroinflammatory activity in mouse BV2 cells
Catalog No. 50-004-39886 Supplier Medchemexpress LLC Supplier No. 250MGHY113301
Hexacosanoic acid is a very long-chain fatty acid. Abnormally elevated levels of hexacosanoic acid are closely associated with various diseases, including X-linked adrenoleukodystrophy, adrenomyeloneuropathy, atherosclerosis, and dementia.
  • Very long-chain fatty acid
  • Associated with various diseases
  • Shows antineuroinflammatory activity in mouse BV2 cells