Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexacosanoic acid-d3 | 2692624-17-0 | MFCD34647473 | >98.0% | 399.71 | C26H49D3O2 | 5 MG
SDP

Catalog No. 5000263928
Encompass_Preferred

Hexacosanoic acid-d3 is a deuterium-labeled stable isotope analogue of hexacosanoic acid used as an analytical standard and tracer in lipidomics and mass spectrometry applications. Supplied as a solid for research use, it provides a reliable internal standard for quantitative analysis.

  • Deuterium-labeled internal standard for mass spectrometry
  • Molecular formula C26H49D3O2; molecular weight 399.71
  • CAS number 2692624-17-0
  • High purity (>98%)
  • Powder stable at -20°C for multi-year storage

Catalog No. 50-002-63928 Supplier Medchemexpress LLC Supplier No. HY113301S25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexacosanoic acid-d3 is a deuterium-labeled stable isotope analogue of hexacosanoic acid used as an analytical standard and tracer in lipidomics and mass spectrometry applications. Supplied as a solid for research use, it provides a reliable internal standard for quantitative analysis.

  • Deuterium-labeled internal standard for mass spectrometry
  • Molecular formula C26H49D3O2; molecular weight 399.71
  • CAS number 2692624-17-0
  • High purity (>98%)
  • Powder stable at -20°C for multi-year storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.