Learn More
Medchemexpress LLC Hexacosanoic acid-d4-1 | 1194984-85-4 | 98.0% | 400.71 | 10 MG
SDP
Catalog No. 5000437861
Encompass_Preferred
Hexacosanoic acid-d4 is the deuterium-labeled version of Hexacosanoic acid. It is a very long-chain fatty acid. Abnormally elevated levels of Hexacosanoic acid are closely associated with various diseases, such as X-linked adrenoleukodystrophy, adrenomyeloneuropathy, atherosclerosis, and dementia. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitation during drug development process.
Catalog No. 50-004-37861 Supplier Medchemexpress LLC Supplier No. HY113301S110MG
Hexacosanoic acid-d4 is the deuterium-labeled version of Hexacosanoic acid. It is a very long-chain fatty acid. Abnormally elevated levels of Hexacosanoic acid are closely associated with various diseases, such as X-linked adrenoleukodystrophy, adrenomyeloneuropathy, atherosclerosis, and dementia. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitation during drug development process.