Learn More
Medchemexpress LLC Hexacosanoic acid-d4-1 | 1194984-85-4 | 98.0% | 400.71 g/mol | C26H48D4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY113301S15MG
Encompass_Preferred
Hexacosanoic acid-d4-1 is a deuterium-labeled reference standard of hexacosanoic acid, a 26-carbon very-long-chain fatty acid. It is supplied for analytical and metabolic research as an isotopically labeled internal standard to support quantitative assays and method development. For research use only.
  • Deuterium-labeled internal standard for quantitative analysis by GC- or LC-MS.
  • High chemical purity: 98.0%.
  • Molecular formula: C26H48D4O2.
  • Available in small pack sizes suitable for analytical use (e.g., 5 MG).
  • Supplied for research use only.
Catalog No. 50-002-63640