Learn More
Cayman Chemical Hexacosanoic Acid methyl ester, Purity: ≥98%, CAS: 5802-82-4, 50 mg
SDP
Catalog No. 502935368
Encompass_Preferred
A fatty acid methyl ester; has been used as a standard for the detection of hexacosanoic acid in beeswax
Catalog No. 50-293-5368 Supplier Cayman Chemical Supplier No. 2672550 MG
A fatty acid methyl ester; has been used as a standard for the detection of hexacosanoic acid in beeswax