Learn More
Cayman Chemical HexacosnoIc AcId 50mg
SDP
Catalog No. 502954991
Encompass_Preferred
A 26-carbon saturated fatty acid; plasma levels are increased in a mouse model of adrenoleukodystrophy; increased levels in red blood cells or in plasma positively correlated with atherosclerotic risk factors and metabolic syndrome, respectively
Catalog No. 50-295-4991 Supplier Cayman Chemical Supplier No. 1335450 MG
A 26-carbon saturated fatty acid; plasma levels are increased in a mouse model of adrenoleukodystrophy; increased levels in red blood cells or in plasma positively correlated with atherosclerotic risk factors and metabolic syndrome, respectively