Learn More
Cayman Chemical Hexadecanal 1g
SDP
Catalog No. 502895153
Encompass_Preferred
A monohydroxy PUFA produced by the actions of 5-LO on a-linolenic acid
Catalog No. 50-289-5153 Supplier Cayman Chemical Supplier No. 90019 1G
A monohydroxy PUFA produced by the actions of 5-LO on a-linolenic acid