Learn More
Cayman Chemical Hexadecanal 5g
SDP
Catalog No. 502894805
Encompass_Preferred
An inhibitor of Spns2 (IC50 94 nM for the mouse transporter) decreases the number of circulating lymphocytes but does not affect plasma S1P levels in mice at 10 mg/kg
Catalog No. 50-289-4805 Supplier Cayman Chemical Supplier No. 90019 5G
An inhibitor of Spns2 (IC50 94 nM for the mouse transporter) decreases the number of circulating lymphocytes but does not affect plasma S1P levels in mice at 10 mg/kg