Learn More
Medchemexpress LLC Hexadecanal | 629-80-1 | 240.42 | 100MG
SDP
Catalog No. 5000448908
Encompass_Preferred
Hexadecanal (Palmitaldehyde) is a volatile long-chain aliphatic aldehyde emitted from human feces, skin, and breath. Its receptor (OR37B) is highly conserved across mammals. It may modulate functional connectivity between brain substrates of social appraisal and aggressive execution. This product is highly deterrent to the ant Lasius niger and shows promise for research into startle responses and aggression.
  • Volatile long-chain aliphatic aldehyde
  • Emitted from human feces, skin, and breath
  • Receptor (OR37B) is highly conserved across mammals
  • May modulate functional connectivity between brain substrates of social appraisal and aggressive execution
  • Highly deterrent to the ant Lasius niger
  • Promising for research on startle responses and aggression
  • Endogenous metabolite
Catalog No. 50-004-48908 Supplier Medchemexpress LLC Supplier No. HYW004305100MG
Hexadecanal (Palmitaldehyde) is a volatile long-chain aliphatic aldehyde emitted from human feces, skin, and breath. Its receptor (OR37B) is highly conserved across mammals. It may modulate functional connectivity between brain substrates of social appraisal and aggressive execution. This product is highly deterrent to the ant Lasius niger and shows promise for research into startle responses and aggression.
  • Volatile long-chain aliphatic aldehyde
  • Emitted from human feces, skin, and breath
  • Receptor (OR37B) is highly conserved across mammals
  • May modulate functional connectivity between brain substrates of social appraisal and aggressive execution
  • Highly deterrent to the ant Lasius niger
  • Promising for research on startle responses and aggression
  • Endogenous metabolite