Learn More
Medchemexpress LLC Hexadecanamide-13,13,14,14,15,15,16,16,16-d9 | 2260669-51-8 | ≥99.0% | 546.96 g/mol | C34H58D9NO3 | 1 MG
SDP
Catalog No. 5000212699
Encompass_Preferred
Deuterium-labeled C16-ceramide (d9) intended as an internal standard and tracer for quantitative analyses by NMR, GC-MS, and LC-MS. The d9 isotopic label enables reliable quantitation of C16-ceramide species in biological samples and analytical workflows. Supplied as a high-purity research chemical in a small, convenient quantity for analytical use.
  • Deuterium-labeled for use as an internal standard.
  • Suitable for NMR, GC-MS, and LC-MS quantitation.
  • High reported purity (≥99%).
  • Appropriate for tracer studies and quantitative assays.
  • Provided in a small analytical quantity for method development.
Catalog No. 50-002-12699 Supplier Medchemexpress LLC Supplier No. HY100354S31MG
Deuterium-labeled C16-ceramide (d9) intended as an internal standard and tracer for quantitative analyses by NMR, GC-MS, and LC-MS. The d9 isotopic label enables reliable quantitation of C16-ceramide species in biological samples and analytical workflows. Supplied as a high-purity research chemical in a small, convenient quantity for analytical use.
  • Deuterium-labeled for use as an internal standard.
  • Suitable for NMR, GC-MS, and LC-MS quantitation.
  • High reported purity (≥99%).
  • Appropriate for tracer studies and quantitative assays.
  • Provided in a small analytical quantity for method development.