Learn More
Medchemexpress LLC Hexadecanamide-13,13,14,14,15,15,16,16,16-d9, N-[(1S,2S,3R)-2,3-dihydroxy-1-(hydroxymethyl)] | 2260669-53-0 | 99.0% | 564.97 | C34H60D9NO4 | 5 MG
SDP
Catalog No. 5000222738
Encompass_Preferred
CER3-d9 is a deuterium-labeled analogue of CER3 supplied for research use as a tracer or internal standard in quantitative analyses. Supplied as a solid in a 5 mg package, it provides high chemical and isotopic purity suitable for analytical workflows including NMR, GC-MS, and LC-MS.
  • Deuterium-labeled analogue useful as tracer or internal standard
  • High purity and isotopic enrichment for accurate quantitation
  • Suitable for NMR, GC-MS, and LC-MS applications
  • Provided as a solid in a small, research-scale package
  • Intended for research use only and not for human use
Catalog No. 50-002-22738 Supplier Medchemexpress LLC Supplier No. HY146831S5MG
CER3-d9 is a deuterium-labeled analogue of CER3 supplied for research use as a tracer or internal standard in quantitative analyses. Supplied as a solid in a 5 mg package, it provides high chemical and isotopic purity suitable for analytical workflows including NMR, GC-MS, and LC-MS.
  • Deuterium-labeled analogue useful as tracer or internal standard
  • High purity and isotopic enrichment for accurate quantitation
  • Suitable for NMR, GC-MS, and LC-MS applications
  • Provided as a solid in a small, research-scale package
  • Intended for research use only and not for human use