Learn More
Apexbio Technology LLC HEXADECANAMIDE-50MG
SDP
Catalog No. 5000570817
Encompass
Hexadecanamide (CAS No 629-54-9) is a bioactive fatty acid amide primarily investigated in the fields of neuroscience and biochemistry Originating from naturally occurring lipids it acts as a modulator of various signaling pathways including the endocannabinoid and fatty acid amide hydrolase systems Its mechanism of action involves interactions with receptors that influence pain inflammation and neuroprotection In vitro its activity is observed in the nanomolar to low micromolar range demonstrating potential therapeutic effects Common applications include studies utilizing cell models such as neuronal and glial cells as well as animal models to explore its effects on pain perception and neurodegenerative diseases It is also used in drug screening for new therapeutic agents targeting similar pathways Experimental concentrations typically depend on the specific research design allowing flexibility in application across different models
Catalog No. 50-005-70817 Supplier Apexbio Technology LLC Supplier No. N288950
Hexadecanamide (CAS No 629-54-9) is a bioactive fatty acid amide primarily investigated in the fields of neuroscience and biochemistry Originating from naturally occurring lipids it acts as a modulator of various signaling pathways including the endocannabinoid and fatty acid amide hydrolase systems Its mechanism of action involves interactions with receptors that influence pain inflammation and neuroprotection In vitro its activity is observed in the nanomolar to low micromolar range demonstrating potential therapeutic effects Common applications include studies utilizing cell models such as neuronal and glial cells as well as animal models to explore its effects on pain perception and neurodegenerative diseases It is also used in drug screening for new therapeutic agents targeting similar pathways Experimental concentrations typically depend on the specific research design allowing flexibility in application across different models