Learn More
Medchemexpress LLC Hexadecanamide, N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]- | 212707-51-2 | MFCD18382130 | 99.0% | 414.62 g/mol | C26H42N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1316785MG
Encompass_Preferred
Palmitoyl serotonin (PA-5HT) is a fatty acid-serotonin hybrid compound that functions as an antagonist of fatty acid amide hydrolase (FAAH). It is supplied as a white to off-white solid with reported purity of 99.0% and is used in preclinical research, notably to study Parkinson's disease and to inhibit L-DOPA-induced dyskinesia in hemi-parkinsonian mouse models. Store powder at -20°C.
  • Acts as an antagonist of fatty acid amide hydrolase (FAAH).
  • Used in preclinical models of Parkinson's disease.
  • Shown to inhibit L-DOPA-induced dyskinesia in animal models.
  • Available as a white to off-white solid with high purity (99.0%).
  • Stable when stored as powder at -20°C for long-term storage.
Catalog No. 50-002-62673