Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexadecanamide (palmitamide) | 629-54-9 | 97.0% | 100 MG
SDP

Catalog No. 5000232523
Encompass_Preferred

Hexadecanamide is a fatty acid primary amide (palmitamide) used in research for its reported anti-inflammatory, antioxidant, and neuroprotective properties. It is supplied as a purified solid for laboratory research; refer to the certificate of analysis and safety data sheet for handling and storage information.

  • Chemical formula: C16H33NO.
  • Molecular weight: 255.44 g/mol.
  • Purity: 97.0% as supplied.
  • Suitable for laboratory research applications.
  • Available in a 100 mg pack.

Catalog No. 50-002-32523 Supplier Medchemexpress LLC Supplier No. HY121389100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexadecanamide is a fatty acid primary amide (palmitamide) used in research for its reported anti-inflammatory, antioxidant, and neuroprotective properties. It is supplied as a purified solid for laboratory research; refer to the certificate of analysis and safety data sheet for handling and storage information.

  • Chemical formula: C16H33NO.
  • Molecular weight: 255.44 g/mol.
  • Purity: 97.0% as supplied.
  • Suitable for laboratory research applications.
  • Available in a 100 mg pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.