Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexadecanamide (palmitamide) | 629-54-9 | 97.0% | 250 MG
SDP

Catalog No. 5000232317
Encompass_Preferred

Hexadecanamide (palmitamide) is a long-chain fatty acid amide used as a research reagent. It has reported anti-allergic, antioxidant, and neuroprotective activities and has been studied in models of mastitis and sperm motility.

  • High purity (97.0%).
  • Molecular formula C16H33NO.
  • Molecular weight 255.44 g/mol.
  • Available in multiple pack sizes, including 250 MG.
  • Solid physical form for straightforward handling and storage.
  • Suitable for in vitro and in vivo research on inflammation and reproductive biology.

Catalog No. 50-002-32317 Supplier Medchemexpress LLC Supplier No. HY121389250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hexadecanamide (palmitamide) is a long-chain fatty acid amide used as a research reagent. It has reported anti-allergic, antioxidant, and neuroprotective activities and has been studied in models of mastitis and sperm motility.

  • High purity (97.0%).
  • Molecular formula C16H33NO.
  • Molecular weight 255.44 g/mol.
  • Available in multiple pack sizes, including 250 MG.
  • Solid physical form for straightforward handling and storage.
  • Suitable for in vitro and in vivo research on inflammation and reproductive biology.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.