Learn More
Medchemexpress LLC Hexadecane 100mL
SDP
Catalog No. 5000205443
Encompass_Preferred
Hexadecane (n-Hexadecane) is a saturated hydrocarbon of alkanes or paraffins and can be used as an organic solvent Hexadecane is a component of petroleum belongs to hydrophobic substrates and is a model substance for studying bacterial degradation of hydrophobic compounds[1]
Catalog No. 50-002-05443 Supplier Medchemexpress LLC Supplier No. HYW099580100ML
Hexadecane (n-Hexadecane) is a saturated hydrocarbon of alkanes or paraffins and can be used as an organic solvent Hexadecane is a component of petroleum belongs to hydrophobic substrates and is a model substance for studying bacterial degradation of hydrophobic compounds[1]